accessibility menu, dialog, popup

UserName

CAS RN 101-64-4

H3CONHH2N

CAS RN 101-64-4

IUPAC Name: N1-(4-methoxyphenyl)benzene-1,4-diamine
Synonyms: N1-(4-methoxyphenyl)benzene-1,4-diamine
Molecular Weight (g/mol): 214.27
Molecular Formula: C13H14N2O
InChi Key: RBLUJIWKMSZIMK-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC2=CC=C(N)C=C2)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results