missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ VapLock™ Solvent Collection Systems with 70 mm Rieke 6TPI Cap Size and 20-L Container

VapLock™ Solvent Collection Systems with 70 mm Rieke 6TPI cap size and 20-L Container are a complete solution to safely collect solvent waste from laboratory instruments

$1,650.06 - $1,888.94

Specifications

For Use With (Equipment) Chromatography Systems
Type Complete Solvent Collection System
Solvent Channels 9
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
13311269
View Documents Promotion Details
Cole-Parmer™
1202067
Polypropylene Manifold
Each for $1,650.06
Only null left
 
13311270
View Documents Promotion Details
Cole-Parmer™
1202068
PTFE Manifold
Each for $1,888.94
Only null left
 
Description

Description

Collect solvent waste from up to 9 effluent streams Minimize exposure to harmful chemical vapors

Warranty

None

Specifications

Specifications

Chromatography Systems
Complete Solvent Collection System
9
Videos
SDS
Documents

Documents