missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanillin, NF, 97-103%, Spectrum™ Chemical
Description
Spectrum™ Chemical Vanillin, NF is used as a flavoring agent in pharmaceutical formulations. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Ignition Residue | 0.0005 |
| Loss on Drying | 0.01 |
| Melting Range | 81-83°C |
| Packaging | Fiber Drum |
| Quantity | 12 kg |
| Percent Purity | 97 to 103% |
| Grade | NF |