missing translation for 'onlineSavingsMsg'
Learn More

Vanillin melting point standard Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012767
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012767 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012767 Supplier Merck Emd Millipore Supplier No. PHR10911G
Only null left

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards

Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 121-33-5
Vapor Pressure >0.01 mmHg (25°C)
Vapor Density 5.3 (vs. air)
Flash Point 153°C - Closed Cup
Physical Form Neat
Linear Formula 4-(HO)C6H3-3-(OCH3)CHO
MDL Number MFCD00006942
Melting Point ∼82°C; 81°C to 83°C
Boiling Point 170°C (15 mmHg)
Synonym Vanillin; 4-Hydroxy-3-methoxybenzaldehyde; Vanillic aldehyde
Type Certified Reference Material
Quantity 1 g
Formula Weight 152.15
Grade Certified Reference Material
Show More Show Less