missing translation for 'onlineSavingsMsg'
Learn More

Vanillin (contains H2SO4) Ethanol Solution [for TLC Stain], TCI America™

Catalog No. v0080500ml
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0080500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V0080500ML Supplier TCI America Supplier No. V0080500ML

Chemical Identifiers

CAS 121-33-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00006942,MFCD08702848
InChI Key MWOOGOJBHIARFG-UHFFFAOYSA-N
Synonym vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
PubChem CID 1183
ChEBI CHEBI:18346
IUPAC Name 4-hydroxy-3-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1O

Specifications

Color Yellow
Quantity 500 mL
UN Number 2924
Formula Weight 152.15
Physical Form Liquid
Chemical Name or Material Vanillin (contains H2SO4) Ethanol Solution [for TLC Stain]
EINECSNumber (3)-1200
RTECSNumber YW5775000
TSCA Yes
Recommended Storage Refrigerator