missing translation for 'onlineSavingsMsg'
Learn More

Vanillin Acetate 98.0+%, TCI America™

Catalog No. v010425g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V010425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V010425G Supplier TCI America Supplier No. V010425G
Only null left

Chemical Identifiers

CAS 881-68-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00003362
InChI Key PZSJOBKRSVRODF-UHFFFAOYSA-N
Synonym vanillin acetate, 4-acetoxy-3-methoxybenzaldehyde, acetovanillin, acetylvanillin, vanillin, acetate, 4-o-acetylvanillin, o-acetylvanillin, benzaldehyde, 4-acetyloxy-3-methoxy, 3-methoxy-4-acetoxybenzaldehyde, acetyl vanillin
PubChem CID 61229
ChEBI CHEBI:86956
IUPAC Name (4-formyl-2-methoxyphenyl) acetate
SMILES CC(=O)OC1=C(C=C(C=C1)C=O)OC

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Vanillin Acetate
EINECSNumber (3)-1204
TSCA Yes