missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VANGL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529088 Supplier Invitrogen™ Supplier No. PA529088
Only null left

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (91%), Rat (91%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Vang-like 1 protein is a novel human homologue of Drosophila Van Gogh/Strabismus. The Vang-like 1 gene consists of at least 7 exons, and encodes a protein with four transmembrane domains and the C-terminal Ser/Thr-X-Val motif. Studies shows that the expression of the protein is significantly up-regulated in gastric cancer cell lines MKN28, MKN74, pancreatic cancer cell lines BxPC-3, PSN-1 and Hs766T. The protein may serve as an effector mediating the intestinal trefoil factor (ITF) healing response of the intestinal mucosa. The protein also serves as a diagnostic marker for hepatocellular carcinoma as well as a potential molecular target for development of novel therapeutic drugs. The pathological role of the protein has been identified in neural tube defects (NTD). Human Vang-like 1 gene maps to chromosome 1p11-p13.1.

Specifications

Antigen VANGL1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene VANGL1
Gene Accession No. Q8TAA9
Gene Alias KAI1 COOH-terminal interacting tetraspanin; KAI1 C-terminal interacting tetraspanin; Kitenin; Loop-tail protein 2; loop-tail protein 2 homolog; LPP2; STB2; Stbm; STBM2; strabismus; strabismus 2; Van Gogh-like protein 1; vang, van gogh-like 1; VANGL planar cell polarity protein 1; Vangl1; vang-like 1 (van gogh, Drosophila); vang-like protein 1
Gene Symbols VANGL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human VANGL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81839
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less