missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vandetanib

Catalog No. AC464332500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464332500 250 mg
AC464330010 1 g
AC464330050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC464332500 Supplier Thermo Scientific Chemicals Supplier No. 464332500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 443913-73-3 | C22H24BrFN4O2 | 475.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 443913-73-3
Molecular Formula C22H24BrFN4O2
Molecular Weight (g/mol) 475.36
InChI Key UHTHHESEBZOYNR-UHFFFAOYSA-N
IUPAC Name N-(4-bromo-2-fluorophenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
SMILES COC1=C(OCC2CCN(C)CC2)C=C2N=CN=C(NC3=CC=C(Br)C=C3F)C2=C1

Specifications

Melting Point 240.0°C to 243.0°C
Solubility Soluble in methanol
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol
Quantity 250 mg
Formula Weight 475.35
Chemical Name or Material Vandetanib

RUO – Research Use Only