missing translation for 'onlineSavingsMsg'
Learn More

Vanadyl Sulfate, Hydrate, 97%, Spectrum™ Chemical

Catalog No. p-7166720
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
20 kg
25 g
5 kg
500 g
Packaging:
Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18614649 25 g Poly Bottle
18614647 100 g Poly Bottle
18614651 500 g Poly Bottle
18614650 5 kg Poly Pail
18614648 20 kg Drum
5 options

Catalog No. 18614649

Supplier: Spectrum Chemical Mfg Cor V102025GM

Only null left

Chemical Identifiers

CAS 123334-20-3

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.