missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadomolybdate Solution, in Dilute Perchloric Acid, LabChem™
$264.57 - $747.23
Specifications
| Chemical Name or Material | Vanadomolybdate Solution |
|---|---|
| Color | Yellow |
| Name Note | in Dilute Perchloric Acid |
| CAS Max % | 88.2,8.8,2,1 |
| CAS | 7601-90-3,12054-85-2,7803-55-6 |
Specifications
| Vanadomolybdate Solution | |
| in Dilute Perchloric Acid | |
| 7601-90-3,12054-85-2,7803-55-6 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| 1235.86 | |
| Poly Bottle | |
| Liquid |
| Yellow | |
| 88.2,8.8,2,1 | |
| (NH4)6Mo7O24·4H2O | |
| Soluble in water | |
| Passes Test | |
| Gaseous Ammonia; Thermal decomposition generates: Corrosive vapors |