missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) oxytriethoxide, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 380010250

Quantity 25mL
Packaging Glass bottle

 View more versions of this product

Catalog No. AC380010250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1686-22-2
202.12
IDIDIJSLBFQEKY-UHFFFAOYSA-N
74312
CCO.CCO.CCO.O=[V]
C6H15O4V
MFCD00799170
triethyl vanadate, vanadium v oxytriethoxide, triethoxyoxovanadium, triethoxo-oxovanadium, vanadium, triethoxyoxo-, t-4, ethylorthovanadate, triethoxy oxo vanadium, iohmvqgdvucilm-uhfffaoysa-n
ethanol;oxovanadium

Specifications

Vanadium(V) oxytriethoxide
1.1390g/mL
71°C
95%
C6H15O4V
(CH3CH2O)3V=O
MFCD00799170
triethyl vanadate, vanadium v oxytriethoxide, triethoxyoxovanadium, triethoxo-oxovanadium, vanadium, triethoxyoxo-, t-4, ethylorthovanadate, triethoxy oxo vanadium, iohmvqgdvucilm-uhfffaoysa-n
CCO.CCO.CCO.O=[V]
202.12
202.12
1686-22-2
125.0°C to 128.0°C
Authentic
Glass bottle
1.5080 to 1.5120
25mL
1.139
IDIDIJSLBFQEKY-UHFFFAOYSA-N
ethanol;oxovanadium
74312
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 216-870-1

SDS
Documents

Documents