missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
Specifications
Specifications
| Chemical Name or Material | Vanadium standard solution |
| Packaging | Plastic bottle |
| Name Note | 1 mg/ml V in 2% HNO3 |
| CAS | 7440-62-2, 7697-37-2, 7732-18-5 |
| Physical Form | Solution |
| Molecular Formula | V |
| Concentration | 995 to 1005 μg/mL of V(3+) in 2% HNO3 (20°C) |
| MDL Number | MFCD00011453 |
| Solubility | Miscible with water |
| InChI Key | LEONUFNNVUYDNQ-UHFFFAOYSA-N |
| Show More |
RUO – Research Use Only