missing translation for 'onlineSavingsMsg'
Learn More

Vanadium Pentoxide, Powder, 99.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor V101012KGBL

 View more versions of this product

Catalog No. 18614640

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1314-62-1
12 kg
Reagent
1
99.5%
Poly Pail

Chemical Identifiers

1314-62-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.