missing translation for 'onlineSavingsMsg'
Learn More

Vanadium Pentoxide, Powder, 99.5%, Spectrum™ Chemical

Catalog No. p-7166719
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614644 25 g Amber Glass Bottle
18614639 125 g Amber Glass Bottle
18614641 500 g Amber Glass Bottle
18614643 2.5 kg Amber Glass Bottle
18614640 12 kg Poly Pail
18614642 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18614644 Supplier Spectrum Chemical Mfg Cor Supplier No. V101025GM
Only null left

Chemical Identifiers

CAS 1314-62-1

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99.5%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.