missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) sulfate oxide hydrate, 99.9% (metals basis)

CAS: 123334-20-3 | O5SV | 163.00 g/mol

Supplier:  Thermo Scientific Chemicals 01109018

 View more versions of this product

Catalog No. AA1109018


Only null left
Explore available promotions

Description

Description

Vanadium(IV) sulfate oxide hydrate is used as a catalyst for the oxidative aromatization of cyclohexenone. It is also used as a component in some food supplements. It is one of the intermediate during the extraction of vanadium from petroleum residues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Vanadium(IV) Sulfate Oxide Hydrate
Lump
99.9%, metal basis
MFCD00149933
Soluble in water.
O=[V++].[O-]S([O-])(=O)=O
163.00
163.00 (Anhydrous)
123334-20-3
50 g
O5SV
Vanadium oxysulfate hydrate
UUUGYDOQQLOJQA-UHFFFAOYSA-L
oxovanadium(2+);sulfate;hydrate
16211496
2.5

Chemical Identifiers

123334-20-3
163.00
UUUGYDOQQLOJQA-UHFFFAOYSA-L
16211496
O5SV
MFCD00149933
Vanadium oxysulfate hydrate
O=[V++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste.

P264b-P270-P301+P312-P330-P501c

H302

DOTInformation : DOT Class: 6.1; Packaging Group: II

EINECSNumber : 248-652-7

RTECSNumber : YW1840000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only