missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) fluoride, 95%

Catalog No. p-7048813
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1154306 5 g
AA1154314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1154306 Supplier Thermo Scientific Chemicals Supplier No. 01154306
Only null left

CAS: 10049-16-8

Vanadium(IV) fluoride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10049-16-8
MDL Number MFCD00042554
Synonym vanadium iv fluoride, acmc-20aja2, vanadium 4+ tetrafluoride, vanadium 4+ ion tetrafluoride, fluoride, fluoride, fluoride, vanadium salt, fluoride

Specifications

Melting Point 325°C (decomposition)
Color Yellow to Brown
Physical Form Powder
Quantity 5 g
Linear Formula VF4
UN Number UN2923
Solubility Information Soluble in water (blue color), acetone (deep green color), and glacial acetic acid (blue-green color)
Formula Weight 126.94
Percent Purity 95%
Sensitivity Moisture sensitive
Density 2.975 g/mL
Chemical Name or Material Vanadium(IV) fluoride
Show More Show Less
Hazard Category H301+H311+H331-H314
Hazard Statement GHS H Statement
H301-H331-H314-H318
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 233-171-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only