Learn More
Vanadium(III) chloride-tetrahydrofuran complex (1:3), 0.5M solution in methylene chl, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 433661000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Vanadium(III) chloride-tetrahydrofuran complex (1:3) | |
| 100mL | |
| VCl·3THF | |
| trichlorotris tetrahydrofuran vanadium, tetrahydrofuran; trichlorovanadium, vanadium iii chloride tetrahydrofuran adduct, vanadium iii chloride tetrahydrofuran complex | |
| C1CCOC1.Cl[V](Cl)Cl | |
| 373.62 | |
| 373.62 | |
| 1.312 |
| 19559-06-9,75-09-2 | |
| VCl3·C12H24O3 | |
| −8°C | |
| WARDLUZTYLPMGJ-UHFFFAOYSA-K | |
| oxolane;trichlorovanadium | |
| 71311352 | |
| AcroSeal™ Glass bottle | |
| 1.3120g/mL |
Chemical Identifiers
| 19559-06-9 | |
| 373.62 | |
| trichlorotris tetrahydrofuran vanadium, tetrahydrofuran; trichlorovanadium, vanadium iii chloride tetrahydrofuran adduct, vanadium iii chloride tetrahydrofuran complex | |
| oxolane;trichlorovanadium |
| VCl3·C12H24O3 | |
| WARDLUZTYLPMGJ-UHFFFAOYSA-K | |
| 71311352 | |
| C1CCOC1.Cl[V](Cl)Cl |
Safety and Handling
GHS P Statement Highly flammable liquid and vapour. Suspected of causing cancer. Causes severe skin burns and eye damage. May cause respiratory irritation. May cause drowsiness or dizziness. May cause damage to organs
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
RUO â Research Use Only