missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valsartan Related Compound B Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 952652-79-8 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C23H27N5O3 |
| MDL Number | MFCD19689078 |
| Synonym | N-(1-Oxobutyl)-N-[[2′-(2 H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-L-valine; N-butyryl-N-{[2′-(1 H-tetrazole-5-yl)-biphenyl-4-yl]methyl}-L-valine; -N-Butyryl-N-{[2′-(1-H-tetrazole-5-yl)-biphenyl-4-yl]methyl}valine |
| Type | Certified Reference Material |
| Quantity | 30 mg |
| Formula Weight | 421.49 |
| Grade | Certified Reference Material |