missing translation for 'onlineSavingsMsg'
Learn More

Valproic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012251
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012251 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012251 Supplier Merck Emd Millipore Supplier No. PHR10611G

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards

Specifications

CAS 99-66-1
Density 0.9 g/mL (at 25°C)
Flash Point 111°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula (CH3 CH2 CH2)2 CHCO2 H
MDL Number MFCD00002672
Boiling Point 220°C
Synonym 2-Propylpentanoic acid; Valproic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 144.21
Grade Certified Reference Material
Show More Show Less