missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valinomycin 93% MP Biomedicals
$232.17 - $576.99
Chemical Identifiers
| CAS | 2001-95-8 |
|---|---|
| Molecular Formula | C54H90N6O18 |
| Molecular Weight (g/mol) | 1111.34 |
| MDL Number | MFCD00005114 |
| InChI Key | FCFNRCROJUBPLU-UYBNATROSA-N |
| Synonym | Cyclo(L-Val-D-HyIva-D-Val-L-Lac-)3: HyIva = α-Hydroxyisovaleric acid; Lac = Lactic acid |
| PubChem CID | 131668508 |
| IUPAC Name | (3S,6S,9R,12R,15S,18S,21R,24R,27S,30S,36R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nonakis(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexaazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone |
| SMILES | CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN10501025
|
N/A | 25 mg |
Each for $232.17
|
|
|||||
|
ICN10501080
|
N/A | 100 mg |
Each for $576.99
|
|
|||||
Chemical Identifiers
| 2001-95-8 | |
| 1111.34 | |
| FCFNRCROJUBPLU-UYBNATROSA-N | |
| 131668508 | |
| CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C |
| C54H90N6O18 | |
| MFCD00005114 | |
| Cyclo(L-Val-D-HyIva-D-Val-L-Lac-)3: HyIva = α-Hydroxyisovaleric acid; Lac = Lactic acid | |
| (3S,6S,9R,12R,15S,18S,21R,24R,27S,30S,36R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nonakis(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexaazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone |
Specifications
| Valinomycin | |
| ≥93% | |
| MFCD00005114 | |
| FCFNRCROJUBPLU-UYBNATROSA-N | |
| (3S,6S,9R,12R,15S,18S,21R,24R,27S,30S,36R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nonakis(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexaazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone | |
| 131668508 | |
| Valinomycin |
| 2001-95-8 | |
| C54H90N6O18 | |
| Cyclo(L-Val-D-HyIva-D-Val-L-Lac-)3: HyIva = α-Hydroxyisovaleric acid; Lac = Lactic acid | |
| CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C | |
| 1111.34 | |
| 1111.4 |
Safety and Handling
EINECSNumber : 217-896-6
Recommended Storage : Store at + 4°C