missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valinomycin 93% MP Biomedicals
Specifications
Specifications
| Type | Valinomycin |
| Physical Form | Powder |
| Color | White |
| Quantity | 100 mg |
| Assay Percent Range | ≥93% |
| Formula Weight | 1111.4 |
| Percent Purity | ≥93% |
| Grade | Not Available |
| Chemical Name or Material | Valinomycin |
Safety and Handling
| EINECSNumber | 217-896-6 |
| Recommended Storage | Store at + 4°C |