missing translation for 'onlineSavingsMsg'
Learn More

Valeryl Chloride 98.0+%, TCI America™

Catalog No. V001025ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V001025ML 25 mL
V0010100ML 100 mL
V0010500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. V001025ML Supplier TCI America Supplier No. V001025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 638-29-9
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000757
InChI Key XGISHOFUAFNYQF-UHFFFAOYSA-N
Synonym valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
PubChem CID 61186
IUPAC Name pentanoyl chloride
SMILES CCCCC(Cl)=O

Specifications

Melting Point -110°C
Color Yellow
Boiling Point 126°C
Quantity 25 mL
UN Number 2502
Formula Weight 120.58
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Valeryl Chloride
EINECSNumber (2)-0632
TSCA Yes