missing translation for 'onlineSavingsMsg'
Learn More

Valeryl chloride, 98%

CAS: 638-29-9 | C5H9ClO | 120.58 g/mol

$108.67 - $108.67

Chemical Identifiers

CAS 638-29-9
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000757
InChI Key XGISHOFUAFNYQF-UHFFFAOYSA-N
Synonym valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
PubChem CID 61186
IUPAC Name pentanoyl chloride
SMILES CCCCC(Cl)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169121000
SDS View Documents
Thermo Scientific Chemicals
169121000
100 g Glass bottle
Each for $108.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

638-29-9
120.58
XGISHOFUAFNYQF-UHFFFAOYSA-N
61186
CCCCC(Cl)=O
C5H9ClO
MFCD00000757
valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
pentanoyl chloride

Specifications

638-29-9
Colorless to Yellow
125.0°C to 127.0°C
Authentic
Glass bottle
1.4190 to 1.4210
MFCD00000757
02,301
valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
XGISHOFUAFNYQF-UHFFFAOYSA-N
pentanoyl chloride
61186
98%
Valeryl chloride, 98%
-110.0°C
0.9900g/mL
32°C
97.5% min. (GC)
C5H9ClO
CH3(CH2)3COCl
100 g
0.99
Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
CCCCC(Cl)=O
120.58
120.58
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously

GHS Signal Word: Danger

EINECSNumber : 211-330-1

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only