missing translation for 'onlineSavingsMsg'
Learn More

Valeric Anhydride 98.0+%, TCI America™

Catalog No. p-7137880
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V000625ML 25 mL
V0006100ML 100 mL
V0006500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. V000625ML Supplier TCI America Supplier No. V000625ML

Chemical Identifiers

CAS 2082-59-9
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.25
MDL Number MFCD00009477
InChI Key DUCKXCGALKOSJF-UHFFFAOYSA-N
Synonym valeric anhydride, n-valeric anhydride, pentanoic anhydride, pentanoic acid, anhydride, n-pentanoic anhydride, pentanoic acid, 1,1'-anhydride, pentanoicacidanhydride, pentanoicacid,anhydride, valeric acid anhydride
PubChem CID 74959
IUPAC Name pentanoyl pentanoate
SMILES CCCCC(=O)OC(=O)CCCC

Specifications

Color Yellow
Boiling Point 227°C
Quantity 25 mL
UN Number 3265
Formula Weight 186.25
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Valeric Anhydride