accessibility menu, dialog, popup

UserName

CAS RN 2082-59-9

IUPAC Name: pentanoyl pentanoate
Synonyms: pentanoic acid, anhydride
Molecular Weight (g/mol): 186.25
Molecular Formula: C10H18O3
InChi Key: DUCKXCGALKOSJF-UHFFFAOYSA-N
SMILES: CCCCC(=O)OC(=O)CCCC
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)