missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valeric Anhydride 98.0+%, TCI America™
Supplier: TCI America V000625ML
Specifications
| Valeric Anhydride | |
| Yellow | |
| C10H18O3 | |
| 25 mL | |
| valeric anhydride, n-valeric anhydride, pentanoic anhydride, pentanoic acid, anhydride, n-pentanoic anhydride, pentanoic acid, 1,1'-anhydride, pentanoicacidanhydride, pentanoicacid,anhydride, valeric acid anhydride | |
| CCCCC(=O)OC(=O)CCCC | |
| 186.25 | |
| 186.25 | |
| Liquid |
| 2082-59-9 | |
| 227°C | |
| MFCD00009477 | |
| 3265 | |
| DUCKXCGALKOSJF-UHFFFAOYSA-N | |
| pentanoyl pentanoate | |
| 74959 | |
| ≥98.0% (T) |
Chemical Identifiers
| 2082-59-9 | |
| 186.25 | |
| DUCKXCGALKOSJF-UHFFFAOYSA-N | |
| 74959 | |
| CCCCC(=O)OC(=O)CCCC |
| C10H18O3 | |
| MFCD00009477 | |
| valeric anhydride, n-valeric anhydride, pentanoic anhydride, pentanoic acid, anhydride, n-pentanoic anhydride, pentanoic acid, 1,1'-anhydride, pentanoicacidanhydride, pentanoicacid,anhydride, valeric acid anhydride | |
| pentanoyl pentanoate |
Safety and Handling
TSCA : Yes