missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valeric Acid 98.0+%, TCI America™
$49.36 - $102.52
Chemical Identifiers
| CAS | 109-52-4 |
|---|---|
| Molecular Formula | C5H10O2 |
| Molecular Weight (g/mol) | 102.133 |
| MDL Number | MFCD00004413 |
| InChI Key | NQPDZGIKBAWPEJ-UHFFFAOYSA-N |
| Synonym | valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n |
| PubChem CID | 7991 |
| ChEBI | CHEBI:17418 |
| IUPAC Name | pentanoic acid |
| SMILES | CCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
V000325ML
|
N/A | 25 mL |
Each for $49.36
|
|
|||||
|
V0003500ML
|
N/A | 500 mL |
Each for $102.52
|
|
|||||
Chemical Identifiers
| C5H10O2 | |
| MFCD00004413 | |
| valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n | |
| CHEBI:17418 | |
| CCCCC(=O)O |
Specifications
| 109-52-4 | |
| Yellow | |
| C5H10O2 | |
| 3265 | |
| NQPDZGIKBAWPEJ-UHFFFAOYSA-N | |
| pentanoic acid | |
| 7991 | |
| 102.13 | |
| Liquid |
| -18°C | |
| 186°C | |
| MFCD00004413 | |
| valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n | |
| CCCCC(=O)O | |
| 102.133 | |
| CHEBI:17418 | |
| ≥98.0% (GC,T) | |
| Valeric Acid |
Safety and Handling
EINECSNumber : (2)-0604
RTECSNumber : YV6100000
TSCA : Yes