missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valeraldehyde 95.0+%, TCI America™
Supplier: TCI America V0001500ML
Specifications
| Valeraldehyde | |
| Yellow | |
| C5H10O | |
| 500 mL | |
| valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde | |
| CCCCC=O | |
| 86.13 | |
| CHEBI:84069 | |
| ≥95.0% (GC) |
| 110-62-3 | |
| 102°C | |
| MFCD00007026 | |
| 2058 | |
| HGBOYTHUEUWSSQ-UHFFFAOYSA-N | |
| pentanal | |
| 8063 | |
| 86.13 | |
| Liquid |
Chemical Identifiers
| 110-62-3 | |
| 86.13 | |
| HGBOYTHUEUWSSQ-UHFFFAOYSA-N | |
| 8063 | |
| pentanal |
| C5H10O | |
| MFCD00007026 | |
| valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde | |
| CHEBI:84069 | |
| CCCCC=O |
Safety and Handling
EINECSNumber : (2)-0488&(2)-0494
RTECSNumber : YV3600000
TSCA : Yes