missing translation for 'onlineSavingsMsg'
Learn More

Valeraldehyde, 97%

Catalog No. p-211639
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149561000 100 mL Glass bottle
AC149562500 250 mL Glass bottle
AC149560010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC149561000 Supplier Thermo Scientific Chemicals Supplier No. 149561000
Only null left

CAS: 110-62-3 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-62-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00007026
InChI Key HGBOYTHUEUWSSQ-UHFFFAOYSA-N
Synonym valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde
PubChem CID 8063
ChEBI CHEBI:84069
IUPAC Name pentanal
SMILES CCCCC=O

Specifications

Melting Point -92.0°C
Density 0.8070g/mL
Boiling Point 103.0°C
Flash Point 12°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3900 to 1.3950
Linear Formula CH3(CH2)3CHO
Quantity 100 mL
Beilstein 01, 676
Specific Gravity 0.807
Merck Index 15, 10088
Solubility Information Solubility in water: 14g/L (20°C). Other solubilities: soluble in alcohol, ether, propylene glycol and, many organic solvents
Viscosity 0.6 mPa.s (20°C)
Formula Weight 86.13
Percent Purity 97%
Chemical Name or Material Valeraldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 203-784-4
RTECSNumber YV3600000
TSCA TSCA

RUO – Research Use Only