missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valacyclovir Hydrochloride Hydrate 97.0+%, TCI America™
Supplier: TCI America V01111G
Specifications
| Valacyclovir Hydrochloride Hydrate | |
| White | |
| MFCD01861507 | |
| L-Valine 2-(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-ylmethoxy)ethyl Ester Hydrochloride | |
| Cl.CC(C)[C@H](N)C(=O)OCCOCN1C=NC2=C1N=C(N)NC2=O | |
| 360.80 | |
| CHEBI:9919 | |
| ≥97.0% (HPLC,N) |
| 124832-27-5 | |
| C13H21ClN6O4 | |
| 1 g | |
| ZCDDBUOENGJMLV-VGVXOIKUNA-N | |
| 2-[(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)methoxy]ethyl (2S)-2-amino-3-methylbutanoate hydrochloride | |
| 67201313 | |
| 360.80 | |
| Crystalline Powder |
Chemical Identifiers
| 124832-27-5 | |
| 360.80 | |
| ZCDDBUOENGJMLV-VGVXOIKUNA-N | |
| 67201313 | |
| 2-[(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)methoxy]ethyl (2S)-2-amino-3-methylbutanoate hydrochloride |
| C13H21ClN6O4 | |
| MFCD01861507 | |
| L-Valine 2-(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-ylmethoxy)ethyl Ester Hydrochloride | |
| CHEBI:9919 | |
| Cl.CC(C)[C@H](N)C(=O)OCCOCN1C=NC2=C1N=C(N)NC2=O |
Safety and Handling
TSCA : No