missing translation for 'onlineSavingsMsg'
Learn More

Valacyclovir Hydrochloride Hydrate 97.0+%, TCI America™

Catalog No. p-7137926
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0111100MG 100 mg
V01111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0111100MG Supplier TCI America Supplier No. V0111100MG
Only null left

Chemical Identifiers

CAS 124832-27-5
Molecular Formula C13H21ClN6O4
Molecular Weight (g/mol) 360.80
MDL Number MFCD01861507
InChI Key ZCDDBUOENGJMLV-VGVXOIKUNA-N
Synonym L-Valine 2-(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-ylmethoxy)ethyl Ester Hydrochloride
PubChem CID 67201313
ChEBI CHEBI:9919
IUPAC Name 2-[(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)methoxy]ethyl (2S)-2-amino-3-methylbutanoate hydrochloride
SMILES Cl.CC(C)[C@H](N)C(=O)OCCOCN1C=NC2=C1N=C(N)NC2=O

Specifications

Color White
Quantity 100 mg
Formula Weight 360.80
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Valacyclovir Hydrochloride Hydrate