missing translation for 'onlineSavingsMsg'
Learn More

Valacyclovir HCl Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012837
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012837 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012837 Supplier Merck Emd Millipore Supplier No. PHR16011G
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Valacyclovir HCl is the hydrochloride salt of L-valyl ester, a prodrug of the antiviral drug acyclovir. Its mode of action involves the inhibition of the viral DNA polymerase and the termination of the viral DNA chain.It is widely used against herpes zoster, herpes simplex and herpes B.

Specifications

CAS 124832-27-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C13H20N6O4 · HCl
MDL Number MFCD01861507
Synonym Valacyclovir hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 360.80
Grade Certified Reference Material