missing translation for 'onlineSavingsMsg'
Learn More

VAI SMA™ Microportable Viable Air Sampler

Catalog No. p-3765906
Change view
Click to view available options
Flow Rate:
28.3, 56.6, and 141.5 L/min.
Weight (Metric):
4.54 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Weight (Metric)
19546936 28.3, 56.6, and 141.5 L/min. 4.54 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19546936 Supplier VAI Supplier No. SMAP40103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Veltek battery charger

The SMA™ Microportable Viable Air Sampler provides consistent environmental monitoring for viable particles.

Battery powered air samplers are intended for use in pharmaceutical clean rooms. The portables draw a measured volume of room air over a sampling plate placed in locations that are critical for environmental quality control sample points.
  • Stainless-steel casing and soft touch controls
  • Rechargeable battery, 8 hours continuous operation
  • Factory-calibrated
  • Plates used are TSA agar type

Warranty

1 Year

Specifications

Material Stainless Steel (316L )
Weight (English) 10 lb.
Product Type Microportable Air Sampler
Flow Rate 28.3, 56.6, and 141.5 L/min.
For Use With (Equipment) 32 mL Media Plates
Weight (Metric) 4.54 kg
Overall Width (English) 7.375 in.
Overall Width (Metric) 18.732 cm
Display Touchscreen