Learn More
BINDER™ Vacuum Pumps VAP with Pump Module
Combined set of vacuum pump and vacuum module including all connection parts.
Supplier: BINDER™ 80121962
Description
- Choice of chemistry diaphragm pumps that are optimally matched to the vacuum ovens.
- Scoop is resistant to a variety of chemicals, especially acids
- Final vacuum can be regulated via the vacuum oven
- Pump module lifts the vacuum oven to the optimum working height, significantly reduces the noise of the vacuum pump, and protects the pump and glass material from mechanical damage.
- Pump in the module can be switched on and off using a switchable socket via the control of the vacuum furnace.
Chemical Identifiers
| Steel | |
| Pump module | |
| 50/60 Hz | |
| 120 V |
Specifications
| Model VD 56 | |
| VAP 1 | |
| 50/60 Hz |
| Steel | |
| Pump module | |
| 120 V |