Learn More
Synthware™ Vacuum and Inert Gas Manifold with Solid Glass Stopcocks, Front Right, Rear Left Connection
GREENER_CHOICE
Catalog No. 31501766
Change view
Click to view available options
No. of Ports:
4
Catalog No. No. of Ports
31501766 4
1 options
Catalog No. 31501766 Supplier Synthware™ Supplier No. M240004
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stainless Steel Retention Clips to ensure stopcocks stay in place.
High vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Stopcocks has constant pressure retention devices to keep them in place. -Hose connection : Back Beam-Left Only, Front Beam-Right only
Specifications
Description Front Right; Rear Left Hose Connection
Type High Vacuum Manifold
For Use With (Application) Organic Synthesis
Includes Solid Glass Stopcocks
Material Borosilicate Glass
No. of Ports 4
Width (Metric) 375 mm