Learn More
Synthware™ Vacuum and Inert Gas Manifold with Hollow Glass Stopcocks, Front Left/Right, Rear Right Spcherical Connection
Description
High vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels. -Hose connection : Back Beam- None, Front Beam- None - 35/20 Socket on right side of back beam and both sides of front beam.
Specifications
Specifications
| Description | Front Left and Right; Rear Right Spherical Connection |
| Type | Spherical Joint Double Manifold |
| For Use With (Application) | Organic Synthesis |
| Includes | Hollow Glass Stopcocks |
| Material | Borosilicate Glass |
| No. of Ports | 4 |
| Width (Metric) | 365 mm |