Learn More
Synthware™ Vacuum and Inert Gas Manifold with Solid Glass Stopcocks, Front Right, Rear Left/Right Connection
GREENER_CHOICE
Catalog No. 31501774
Change view
Click to view available options
No. of Ports:
5
Catalog No. No. of Ports
31501774 5
1 options
Catalog No. 31501774 Supplier Synthware™ Supplier No. M300005
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stainless Steel Retention Clips to ensure stopcocks stay in place.
High vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Stopcocks has constant pressure retention devices to keep them in place. -Hose connection : Back Beam-both, Front Beam- Right only
Specifications
Description Front Right; Rear Left and Right Hose Connection
Type High Vacuum Manifold
For Use With (Application) Organic Synthesis
Includes Solid Glass Stopcocks
Material Borosilicate Glass
No. of Ports 5
Width (Metric) 450 mm