Learn More
Synthware™ Vacuum or Inert Gas Manifold, Double, Front Left, Rear Left/Right Connection
GREENER_CHOICE
Catalog No. 31501735
Change view
Click to view available options
No. of Ports:
5
Catalog No. No. of Ports
31501735 5
1 options
Catalog No. 31501735 Supplier Synthware™ Supplier No. M171005
General purpose double bank manifold for use with vacuum and inert gas.
Supplied with 0 to 4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm.
Specifications
Connection Type Front Left; Rear Left and Right Hose Connection
Type Vacuum/Inert Gas Manifold
For Use With (Application) Organic Synthesis
Includes High Vacuum Valve Plugs
Material Borosilicate Glass
No. of Ports 5
Width (Metric) 550 mm