Learn More
Pyramex™ V2G-Plus™ Safety Eyewear
H2X anti-fog dual lenses
Supplier: Pyramex™ GB6410SDT
Description
Spécifications
| Anti-fog | |
| ANSI Z87.1,CSA Z94.3-07 | |
| Clear | |
| Black | |
| 80 g | |
| TPU |
| H2X | |
| EN-166 | |
| Goggles | |
| Goggle with Strap | |
| Polycarbonate (Front Lens), Acetate (Inner Lens), PU Foam |