missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP Scientific Freestanding / Mobile Glassware Washers
Designed for ease of operation and versatile loading, SP Hotpack Freestanding / Mobile Laboratory Glassware Washers include sturdy casters that allow flexible placement in the laboratory. Equipped with premium features including a soft touch controller, enhanced drying, and quiet run cycles, SP Hotpack Laboratory Glassware Washers are engineered for durability and energy efficient operation.
$34,999.46 - $41,853.75
Specifications
| Display | Digital |
|---|---|
| Material | Stainless Steel |
| Max. Pressure | 60 psi |
| Water Consumption | Min: Tap 2.4 gal. (9L), DI 0.8 gal. (3L); Max: Tap 4 gal. (15L), DI 1.6 gal. (6L) |
| Certifications/Compliance | MET Labs |
| Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
|
15365213
|
SP Scientific
UCWHA10101 |
DI Rinse - Top and Bottom Standard Racks |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
15365214
|
SP Scientific
UCWHA20101 |
DI Rinse - Top and Bottom Spindle Rack Ready |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Provide reliable day-in, day-out performance so your laboratory glass is always ready-to-go
- All models come with removable top and bottom stainless-steel open racks to accommodate a variety of glassware
- Spindle rack ready models accommodate direct injection spindle racks (sold separately) for thorough cleaning inside narrow-neck glassware
- Stainless steel baskets, insert racks and accessories sold separately to maximize space, improve efficiency, and protect specialty glassware
- Temperature selection to 70°C (158°F); can effectively dissolve stubborn oils
- DI rinse models ensure a spotless finish; easily connect to fixed or portable DI water systems
- Optional DI Booster Pump recommended when the incoming DI water source has a pressure of less than 18psi
- Delayed start feature enables washer to operate unattended during off-peak hours to reduce energy costs
- Designed for optimal quiet operation with noise buffering insulation
- Three filters protect pumps from debris; conveniently located for easy access
- Minimal, efficient water consumption
- Enhanced dual fan drying system for spotless results
- 304 stainless steel chamber and exterior casing ensures corrosion protection and long life
- 3 in. lockable casters allow easy, controlled mobility of unit
- UL 61010-1 and CSA 22.2 61010-1 certified by MET Labs
- Microprocessor control system features 6 cycle selections including a customizable timed option to ensure minimum energy consumption. All wash cycles have pre-wash, wash, pre-rinse and rinse cycles to prevent contamination.
- Rinse Only - Short tap water rinse cycle
- Normal Wash - Lower temperature (55°C) longer cycle with three (3) final rinse cycles
- Daily Wash - Standard temperature (65°C) with two (2) final rinse cycles
- Heavy Wash - Higher temperature (70°C) longer cycle with multiple pre-wash and final rinse cycles
- Heavy Wash Time Saver - Lower temperature (55°C) version of Heavy Wash with a shorter overall cycle time
- Timed Wash Programs - Standard temperature (65°C) in programmable 15 minute increments up to 2.5 hours
Specifications
| Digital | |
| 60 psi | |
| MET Labs | |
| 55°-70°C (131°-158°F) | |
| 61.6 cm | |
| 97.8 cm | |
| 120V, 60Hz, 15A |
| Stainless Steel | |
| Min: Tap 2.4 gal. (9L), DI 0.8 gal. (3L); Max: Tap 4 gal. (15L), DI 1.6 gal. (6L) | |
| Washing Lab Glassware | |
| 24.25 in. | |
| 38.5 in. | |
| 62 kg | |
| 60 Hz |