Learn More
UVP White Light Transilluminator
Description
- Double-safety sealed plexiglass and glass surfaces
- Scratch and chemical resistant work area
- Uniform light diffusion
- Film clip for stability
Specifications
Specifications
| Product Type | White Light Transilluminator |
| Includes | Plexiglass and glass work surface for durability; Scratch/chemical resistant work area; Edge clip on filter glass to hold film |
| Filtration Area (Metric) | 36 x 43 cm |
| Electrical Requirements | 115 V |
| Size (Metric) Filter | 36 x 43 cm |