Learn More
UVP PhotoDoc-It™ Imaging Systems
Economical and compact photodocumentation systems capture color images for basic gel documentation
Supplier:  UVP 97027404
 View more versions of this product
Catalog No. UVP97027404
Only null left
Explore available promotions
Includes: Digi Camera, PhotoDoc-It hood, Ethidium Bromide filter, color printer; UV transilluminator (except UVP97027401), combination lock
Description
Description
  • Contains high resolution megapixel color camera
  • Compact, lightweight hood enclosure creates a darkroom environment
  • Combination lock secures camera to the hood
  • UV transilluminators contain single or multiple wavelengths and 20 × 20cm or 21 × 26cm filter sizes
  • Generate lab quality 4 x 6 in. prints with mini color printer provided, includes 108 sheets
  • Can be connected to a computer for transfer of images
  • View gels through the gel viewer window
Specifications
Specifications
Translluminator M-20
Yes
High resolution megapixel digital color camera
Imaging System
Product Suggestions
Product Suggestions