Learn More
UVP PhotoDoc-It™ Imaging Systems
Catalog No. UVP97027413
Change view
Click to view available options
Transilluminator:
Translluminator LM-20E
Translluminator LM-26E
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Transilluminator
UVP97027413 Translluminator LM-26E
UVP97027407 Translluminator LM-20E
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. UVP97027413 Supplier UVP Supplier No. 97027413
Includes
Digi Camera, PhotoDoc-It hood, Ethidium Bromide filter, color printer; UV transilluminator (except UVP97027401), combination lock
Economical and compact photodocumentation systems capture color images for basic gel documentation
  • Contains high resolution megapixel color camera
  • Compact, lightweight hood enclosure creates a darkroom environment
  • Combination lock secures camera to the hood
  • UV transilluminators contain single or multiple wavelengths and 20 × 20cm or 21 × 26cm filter sizes
  • Generate lab quality 4 x 6 in. prints with mini color printer provided, includes 108 sheets
  • Can be connected to a computer for transfer of images
  • View gels through the gel viewer window
Specifications
Transilluminator Translluminator LM-26E
Printer Included Yes
Camera Type High resolution megapixel digital color camera
Product Type Imaging System