Learn More
Analytik Jena™ UVP EL Series Lamps
Select from lamps with four, six or eight watts.
Supplier: Analytik Jena™ 95027401
Description
- Manufactured with a durable, extruded metal housing and scratch resistant powder paint.
- Models available include 254nm shortwave, 365nm longwave or a combination UV wavelengths or UV and white light versions.
- Basic configuration offers one UV and one white light tube.
- Twin models have two tubes of the same wavelength or the 2UV™ Models use two different wavelengths.
- The 3UV™ Model available only in the eight watt lamp, gives three UV wavelengths in one lamp! No other lamp offers this versatility. Easily switch among the three wavelengths (longwave, shortwave and midrange UV) as your application requires.
- Can be placed in the J-138 stand for hands-free use.
Applications:
Bacterial identification, specimen staining, gel electrophoresis, optical lab measurements, fluorescence demonstration, biochemical testing
Specifications
| 6 W | |
| Metal | |
| 6 W, 115 V, 60 Hz | |
| UVP UVS-16 Lamp |
| 254 nm | |
| 12 x 3.8 in. | |
| Bacterial identification, specimen staining, Gel electrophoresis, optical lab measurements, fluorescence demonstration, biochemical testing | |
| 1 Ea. |