Learn More
UVP UV Light Converter Plates
Maximize transilluminator capabilities by using a converter plate to convert the transilluminator's 302nm UV wavelength to different wavelengths for visualizing a variety of gels. Place the specially designed plate on the transilluminator filter surface.
Supplier:  UVP 38020001
 View more versions of this product
Catalog No. UVP38020001
Only null left
Description
Description
  • Convert UV to 460-470nm with the Visi-Blue™ plate for viewing GFP stains
  • Scratch-resistant glass is mounted in a metal housing for durability
  • Rubber seal stops the plate from sliding
  • Handles are situated on two sides of the plate for easy handling
  • Several standard plate sizes are available
  • Variety of wavelengths for gel documentation, gel excitation and gel imaging
Specifications
Chemical Identifiers
Converter Plate
Specifications
Converter Plate
8.26 x 10.23 in.
Visi-Blue Converter Plates
Convert 302nm UV to 460 to 470nm Blue Light
21 x 26 cm
Product Suggestions
Product Suggestions