Learn More
UVP UV Light Converter Plates
Maximize transilluminator capabilities by using a converter plate to convert the transilluminator's 302nm UV wavelength to different wavelengths for visualizing a variety of gels. Place the specially designed plate on the transilluminator filter surface.
Supplier:  UVP 38019104
 View more versions of this product
Catalog No. 38019104
Only null left
Description
Description
  • White light plates convert UV to white light for viewing protein gels, coomassie blue stained or silver stained media; uniquely phosphored glass assembly (patent pending) converts the UV radiation via a white diffuser
  • Scratch-resistant glass is mounted in a metal housing for durability
  • Rubber seal stops the plate from sliding
  • Handles are situated on two sides of the plate for easy handling
  • Several standard plate sizes are available
  • Variety of wavelengths for gel documentation, gel excitation and gel imaging
Specifications
Chemical Identifiers
Converter Plate
Specifications
Converter Plate
9.8 x 10.2 in.
UV to White Light Converter Plates
Convert 302nm UV to White Light
25 x 26 cm
Product Suggestions
Product Suggestions