missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UTS2 Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is >95% (by SDS-PAGE).
Urotensin II is an active cyclic heptapeptide absolutely conserved from lamprey to human. The active peptide acts as a vasoconstrictor and is expressed only in brain tissue. Despite the gene family name similarity, this gene is not homologous to urocortin, a member of the sauvagine/corticotropin releasing factor/urotensin I family. Most of the proprotein is cleaved to make the mature peptide.
Specifications
Specifications
| Antigen | UTS2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | UTS2 |
| Gene Accession No. | O95399, Q9QZQ3 |
| Gene Alias | prepro U-II; prepro-UII; PRO1068; Ucn2; UII; U-II; UNQ525/PRO1068; urocortin 2; urotensin 2; urotensin II; Urotensin-2; urotensin-II; UTS2 |
| Gene Symbols | UTS2 |
| Show More |