Learn More
BIOTIX™ uTIP™ Non-Sterile Low Retention Universal Pipette Tips, Standard
Designed with FlexFit™ to lower necessary insertion and ejection force by ≈70% to make secure seal. Non-Filter Pipette Tips for Universal Pipettes are manufactured with low retention X-Resin™ giving tighter CV values.
$505.33 - $505.33
Specifications
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model Universal
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
12-111-128
View Documents Promotion Details
BIOTIX™
63300058
250 μL
Case of 4800 Each for $505.33
Only null left
12-111-134
View Documents Promotion Details
BIOTIX™
63300060
1000 μL
Case of 3840 Each for $505.33
Only null left
Description
Description
  • Compatible with the widest range of industry leading pipettes - Fits all standard pipettes
  • Accurate and precise channel-channel draw on multichannel pipettes
  • Secure seal on single and multichannel models
  • FlexFit™ seals on your pipette with less force, reducing your ejection force
  • FlexFit™ technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
  • X-Resin™ maximizes your sample recovery by creating a low binding surface for DNA, proteins, and nanoparticles - resulting in tighter CV values
  • Blade™ lowers chance of hanging droplet formation by optimizing surface area at distal end of tip - ensures a clean delivery of your sample, increasing accuracy and precision
  • Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
  • Better reprodicibility
  • Certifications
    ISO 9001
    Specifications
    Specifications
    Non-filtered
    Clear
    Universal
    Reusable as a 96 well reservoir
    Hinged Rack
    Racked
    Pipette Tips
    Universal Fit
    uTip
    Conical
    Non-sterile
    Autoclavable
    ISO 9001
    Low binding for Protein, DNA, and nanoparticles
    Ergonomic
    10 racks/pk
    Single-use
    Yes
    Polypropylene
    Videos
    SDS
    missing translation for 'documents'
    missing translation for 'documents'