Learn More
BIOTIX™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk
Top quality pipette tips in resealable bags. Validated for fit and accuracy on universal pipettes.
$503.57 - $503.57
Specifications
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Not Autoclavable
Model Universal
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Price Qty  
Catalog Number Mfr. No. Volume (Metric) Price Qty  
12-111-131
View Documents Promotion Details
BIOTIX™
63300079
250 μL
Case of 10000 Each for $503.57
Only null left
12-111-137
View Documents Promotion Details
BIOTIX™
63300081
1000 μL
Case of 4000 Each for $503.57
Only null left
Description
Description
  • Validated for fit and accuracy on standard pipettes like Eppendorf™, Gilson™ and Biohit™
  • Low binding for Protein, DNA and nanoparticles
  • Re-sealable bag
  • Tips must be racked by hand
  • Certifications
    ISO 9001
    Specifications
    Specifications
    Non-filtered
    Clear
    Universal
    Fully Recyclable
    Bulk
    1000 tips/bag
    Single-use
    Yes
    Polypropylene
    Non-sterile
    Not Autoclavable
    ISO 9001
    Low binding for Protein, DNA, and nanoparticles
    Ergonomic
    Pipette Tips
    Universal Fit
    uTip
    Conical