missing translation for 'onlineSavingsMsg'
Learn More

Calcium Stearate, FCC, 9-10.5%, Spectrum™ Chemical

Catalog No. 18601384
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601384 45 kg Poly Pail
18601381 500 g Poly Pail
18601382 2.5 kg Poly Pail
18601383 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601384 Supplier Spectrum Chemical Mfg Cor Supplier No. C113145KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1131, 1592-23-0

Spectrum™ Chemical Calcium Stearate, FCC , also known as E470, is used as a flow agent in powders. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1592-23-0
Molecular Formula C36H70CaO4
Molecular Weight (g/mol) 607.03
InChI Key CJZGTCYPCWQAJB-UHFFFAOYSA-L
IUPAC Name calcium dioctadecanoate
SMILES [Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

Physical Form Powder, Crystals and/or Chunks
CAS Max % 100%
Quantity 45 kg
Loss on Drying 4%
Packaging Poly Pail
Percent Purity 9 to 10.5%
Grade FCC
Chemical Name or Material Calcium Stearate